8.22334
Hydroxylammonium chloride
for synthesis
Synonym(s):
Hydroxylammonium chloride, Hydroxylamine hydrochloride
About This Item
vapor pressure
0.054 Pa ( 50 °C)
Quality Level
Assay
≥99% (argentometric)
form
solid
pH
2.5-3.5 (20 °C, 50 g/L in H2O)
mp
154 °C
solubility
470 g/L
density
1.70 g/cm3 at 20.2 °C
bulk density
900 kg/m3
storage temp.
15-25°C
SMILES string
[Cl-].NO.[H+]
InChI
1S/ClH.H3NO/c;1-2/h1H;2H,1H2
InChI key
WTDHULULXKLSOZ-UHFFFAOYSA-N
Analysis Note
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral
Target Organs
spleen
Storage Class Code
4.1A - Other explosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service