Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81516

Supelco

Poly(α-methylstyrene)

analytical standard, 10,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(C6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~7800
Mp ~8500
Mw ~8300

technique(s)

gel permeation chromatography (GPC): suitable

density

1.075 g/mL at 25 °C (lit.)

Mw/Mn

~1.07

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)c1ccccc1

InChI

1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3

InChI key

XYLMUPLGERFSHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(α-methylstyrene) may find applications as a radical producing agent, in the thermal degradation of polystyrene.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal degradation of vinyl polymers I?Thermal degradation of polystyrene-poly (?-methylstyrene) mixtures
Richards.H.D and Salter.A.D
Polymer, 8, 127-138 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service