Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15355

Sigma-Aldrich

N-Boc-ethanolamine

purum, ≥96.0% (GC)

Synonym(s):

N-(tert-Butoxycarbonyl)ethanolamine, tert-Butyl N-(2-hydroxyethyl)carbamate, Boc-2-aminoethanol, Boc-glycinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHCH2CH2OH
CAS Number:
Molecular Weight:
161.20
Beilstein:
2353995
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

purum

Quality Level

Assay

≥96.0% (GC)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.449 (lit.)
n20/D 1.450

density

1.042 g/mL at 25 °C (lit.)

SMILES string

OCCNC(OC(C)(C)C)=O

InChI

1S/C7H15NO3/c1-7(2,3)11-6(10)8-4-5-9/h9H,4-5H2,1-3H3,(H,8,10)

InChI key

GPTXCAZYUMDUMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Building block for phospholipids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.-L. Alcaraz et al.
The Journal of Organic Chemistry, 61, 192-192 (1996)
S.F. Martin et al.
Tetrahedron Letters, 29, 3631-3631 (1988)
S.F. Martin et al.
The Journal of Organic Chemistry, 59, 4805-4805 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service