XX6320230
Power Supply, 230 V
fecal coliforms, fecal streptococci, microorganisms
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104424
NACRES:
NB.11
Recommended Products
AC/DC input
230 V, 50 Hz
Quality Level
application(s)
cosmetics
environmental
food and beverages
microbiology
shipped in
ambient
General description
230 V rechargeable nickel-cadmium battery
Application
fecal coliforms, fecal streptococci, microorganisms
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service