Recommended Products
type
for cell culture
concentration
(Please refer to lot specific datasheet.)
technique(s)
cell based assay: suitable
detection method
fluorometric
Application
Live cell fluorescent imaging
Quality
Purity: ≥ 98% confirmed by HNMR, LC-MS and HPLC and elemental analysis
Molar Mass: 706.68 g/mol
Molar Mass: 706.68 g/mol
Physical properties
706.68 g/mol
Physical form
Lyophilized
Storage and Stability
Store BioTracker™ Cys Hcy GSH Live Cell Dye at -20°C, desiccate and protect from light Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.
Legal Information
BioTracker is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service