Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RMTSH-200K

Millipore

MILLIPLEX® Rat/Mouse TSH Magnetic Bead Panel

new

Synonym(s):

Luminex TSH immunoassay, Millipore Rat/Mouse TSH panel, Rat/Mouse TSH Panel, Thyroid Stimulating Hormone Assay, thyrotropin assay, thyrotroponin panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

description

Research category : Endocrine
Research sub-category : Metabolism, Metabolic disorders

Quality Level

product line

Milliplex®

species reactivity

rat

assay range

intra-assay cv: ≤10%
standard curve range: 18-40,000 pg/mL
inter-assay cv: ≤20%

technique(s)

multiplexing: suitable using : suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Thyroid Stimulating Hormone (TSH), also referred to as thyrotropin, is vital in controlling the thyroid gland′s function. It prompts the thyroid to produce the essential hormones T3 and T4, which are critical for metabolism. Abnormal TSH levels are linked to thyroid disorders, and TSH is extensively studied as a significant biomarker in endocrine disruptor research. MILLIPLEX® Rat/Mouse TSH Magnetic Bead Panel is a Single-plex kit to be used for the quantification of Thyroid Stimulating Hormone (TSH) in rat and mouse serum, plasma, and tissue culture samples

Specificity

There was no or negligible cross-reactivity.

Application

The MILLIPLEX® Rat/Mouse TSH Magnetic Bead Panel can be utilized to quantify TSH levels for the study of the following conditions:

  • High TSH (Hypothyroidism-Related Disorders)
  • Endocrine Disruptor Studies
  • Low TSH (Hyperthyroidism-Related Disorders)
  • Pituitary or Hypothalamic Disorders (Abnormal TSH levels)
  • Thyroid Cancer
  • Pregnancy-Related Thyroid Dysfunction
  • Chronic Kidney Disease

Features and Benefits

  • TSH Single-plex Assay
  • 8 Point Standard Curve.
  • 3 Quality Controls included.
  • Optimized for Rat, Mouse: Serum, Plasma and Tissue Culture Samples

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service