Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NWKITNC

Millipore

Immobilon® -NC Nitrocelluose NOW Bundle

2 rolls, 8.5 cm x 10 m, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane, 1 dispenser included

Synonym(s):

nitrocellulose transfer membrane, Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NA.32

Product Name

Immobilon®-NC NOW Bundle, 2 rolls, 8.5 cm x 10 m, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane, 1 dispenser included

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

feature

hydrophilic

packaging

pkg of 2 roll dispenser box of 1

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter L × W

8.5 cm × 10 m

pore size

0.45 μm pore size

compatibility

for use with

  • Amido black
  • CPTS
  • Colloidal gold
  • India ink
  • Ponceau-S red

detection method

colorimetric
fluorometric
radioactive

shipped in

ambient

General description

This bundle includes the Immobilon NOW Dispenser and two 8.5cm x 10m rolls of Immobilon-NC transfer membrane. The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7 x 8.5cm) and midi-blots (8.5 x 13.5cm).

Application

Immobilon-NC Transfer Membrane has been used in western blot analysis.

Compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service