NU050/452-312
Valve Body for NOVASEPTIC® Valve, Tank Outlet 180°, Elbow 45°
TC inlet, TC outlet
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151800
Recommended Products
material
device (PFTE, polytetrafluoroethylene)
diaphragm (silicone)
silicone valve body
Quality Level
feature
hydrophobic (Surface)
parameter
-40-170 °C temp. range (continous use)
-40-250 °C temp. range (short term use)
weight
0.44 kg (0.970 lb)
fitting
TC inlet
TC outlet
Application
The Diaphragm is manufactured from 100% solid PTFE bar.
The applied valve body and actuator may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.1550 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service