Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NS350-5ML

Sigma-Aldrich

EZ-Float®Tissue Mounting Reagent 100X

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NA.47

technique(s)

immunohistochemistry: suitable (tissue)

Quality Level

shipped in

ambient

General description

EZ-Float Tissue Mounting Reagent is a novel aqueous solution for mounting free-floating tissue sections onto glass slides. EZ-Float enhances sample quality by minimizing folding or tearing of tissue sections, and reduces your sample processing time 3-4 fold compared to buffers such as PBS or TBS.

Application

Preparation of Reagent
EZ-Float Tissue Mounting Reagent is provided as a 100X stock solution which must be diluted to a 1X working solution using PBS or deionized water immediately prior to use. The 1X working solution should be made fresh for each experiment.

Storage and Stability

Store the 100X EZ-Float Reagent at 2-8°C. When stored properly the 100X solution is stable for 12 months from date of receipt.

Legal Information

EZ-FLOAT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For research and further manufacturing use only. It is not for use in humans.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service