Key Documents
NA25/22TR
Millipore
NovAseptic Valve, Diaphragm, EPDM
About This Item
UNSPSC Code:
31411800
material
EPDM rubber device (ethylene propylene diene monomer rubber)
PTFE
Quality Level
product line
NovAseptic®
feature
hydrophobic (Surface)
parameter
—1 - 6 bar pressure range
—30 - 135 °C temp. range (Short term use)
weight
0,01 kg (0.022 lb)
Application
The NovAseptic® valve is specifically designed for aseptic applications and complies with the most stringent cleanability and sterilization requirements. The focus on aseptic design is a significant feature for all valves. The NovAseptic® valve is designed for minimum dead leg, complete drainability and high chemical resistance.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany