Recommended Products
Product Name
Anti-GAPDH (CT) Antibody, clone RM114,
antibody form
purified antibody
Quality Level
clone
RM114
monoclonal
purified by
using Protein A
species reactivity
human, rabbit
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG
UniProt accession no.
General description
A peptide corresponding to the C-terminus of GAPDH.
Application
Epitope: C-terminus
Quality
Evaluated by Western Blotting in A431 cell lysate.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected GAPDH (CT) in A431 cell lysate.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected GAPDH (CT) in A431 cell lysate.
Physical properties
~39 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Physical form
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service