Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2489

Anti-SCUBE1 Antibody, clone 701

new

Synonym(s):

Signal peptide, CUB and EGF-like domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

701, monoclonal

mol wt

calculated mol wt 107.91 kDa
observed mol wt ~130 kDa

purified by

using protein G

species reactivity

monkey, human, mouse

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... SCUBE1(80274)

Specificity

Clone 701 is a mouse monoclonal antibody that specifically detects SCUBE1 but not, SCUBE2 or SCUBE3. It targets an epitope within 200 amino acids from the C-terminal region.

Immunogen

GST-tagged, recombinant fragment from the C-terminus of human SCUBE1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK-293T cells transfected with pFlag-hSCUBE1.

Western Blotting Analysis: A 1:500 dilution from a representative lot detected SCUBE1 in lysate from HEK-293T cells transfected with pFlag-hSCUBE1.

Tested Applications

Western Blotting Analysis: A representative lot detected SCUBE1 in Western Blotting application (Tu, C-F., et al. (2006). Cardiovasc Res. 71(3):486-95; Dai, D-F, et al. (2008). J Am Coll Cardiol. 51(22):2173-80; Liao, W-J, et al. (2019). Biochim Biophys Acta Mol Basis Dis. 1865(2):329-338).

Immunofluorescence Analysis: A representative lot detected SCUBE1 in Immunofluorescence application (Tu, C-F., et al. (2006). Cardiovasc Res. 71(3):486-95).

Flow Cytometry Analysis: A representative lot detected SCUBE1 in Flow Cytometry application (Tu, C-F., et al. (2006). Cardiovasc Res. 71(3):486-95).

Immunohistochemistry Analysis: A representative lot detected SCUBE1 in Immunohistochemistry application (Tu, C-F., et al. (2006). Cardiovasc Res. 71(3):486-95; Liao, W-J, et al. (2019). Biochim Biophys Acta Mol Basis Dis. 1865(2):329-338).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Signal peptide, CUB and EGF-like domain-containing protein 1 (UniProt: Q8IWY4) is encoded by the SCUBE1 gene (Gene ID: 80274) in human. The gene encodes a platelet-endothelial cell surface protein that is expressed during early embryogenesis. The protein plays a role during the early stages of central nervous system development. SCUBE1 is a novel adhesive molecule that mediates platelet matrix interaction and ristocetin-induced platelet agglutination. It is produced with a signal peptide sequence at the N-terminus (aa 1-23) followed by nine tandemly arranged epidermal growth factor (EGF)-like repeats (aa 33-402), a spacer region and three cysteine-rich repeat motifs and a CUB domain at the C-terminus (aa 798-910). SCUBE1 protein is highly expressed in platelets and stored in the platelet -granules, and translocated to the surface upon platelet stimulation. Studies suggest that matrix-bound and soluble fragments of SCUBE1 may play critical (patho)physiological roles in cardiovascular biology. Platelet activation plays a crucial role in acute coronary syndrome (ACS) and acute ischemic stroke (AIS), and platelet stimulation is associated with increased plasma concentration of SCUBE1. Studies show plasma SCUBE1 concentration is significantly elevated in ACS and AIS but, not in coronary artery disease (CAD) patients. Plasma SCUBE1 is implicated to be a potential biomarker of platelet activation in acute thrombotic disease. It may also function in renal regeneration and repair. (Ref.: Tu, C-F., et al. (2006). Cardiovasc Res. 71(3):486-95; Dai, D-F, et al. (2008). J Am Coll Cardiol. 51(22):2173-80; Liao, W-J, et al. (2019). Biochim Biophys Acta Mol Basis Dis. 1865(2):329-338).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service