Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2181

Sigma-Aldrich

Anti-Phosphotyrosine (Fab fragment) Antibody, clone 4G10-G4

clone 4G10-G4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

4G10-G4, monoclonal

Quality Level

Specificity

Clone 4G10-G4 is a his tagged recombinant Fab fragment that detects phosphotyrosine in proteins from all species. To be used with anti-His Tag secondary antibody.

Immunogen

Recombinant antibody from Fabs constructed in a dual expression vector for Light chain and His-tagged heavy chain of clone 4G10 and expressed in C43(DE3) strain of E. coli.

Reconstitution

1 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service