Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN824

Sigma-Aldrich

Anti-α-Synuclein (SNCA) Antibody

mouse monoclonal, LB509

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-α-syn Antibody (amino acids 115-122), clone LB509,

antibody form

purified antibody

Quality Level

clone

LB509
monoclonal

purified by

using protein G

species reactivity

human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

General description

Conjugated linear peptide corresponding to human α-syn (aa 115-122).

Specificity

This antibody detects a-Synuclein preformed fibrils that can cross seed tau fibrilliation.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected α-syn in human cerebral cortex tissue.
Western Blotting Analysis: A representative lot detected α-syn in PK-digested FL a-syn pffs (Guo, J.L., et al. (2013). Cell. 154:103-117).

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected α-syn in 10 µg of human brain tissue lysate.

Physical properties

~15 kDa observed

Physical form

Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service