MABN2420
Anti-Amyloid β 1-42 Antibody, clone mHJ5.1
clone mHJ5.1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
Recommended Products
Specificity
Clone mHJ5.1 is a mouse monoclonal antibody that specifically detects Aβ 1-42 peptide. It targets an epitope within 12 amino acids from the internal region of Aβ 1-42 peptide. Does not exhibit cross reactivity with other APP fragments.
Immunogen
A linear peptide corresponding to 12 amino acids from the internal region of human Aβ 1-42 peptide
Reconstitution
0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Storage and Stability
Stable for 1 year at +2°C to +8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service