MABF2347
Anti-VSV Matrix Protein M Antibody, clone 23H12
clone 23H12
Synonym(s):
Anti-VSV Matrix Antibody, Clone 23H12 Anti-VSV, VSV Matrix Protein Detection
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
clone
23H12, monoclonal
isotype
IgG2aκ
Specificity
Clone 23H12 is a mouse monoclonal antibody that detects Vesicular stomatitis virus matrix protein M in virus infected cells.
Immunogen
Vesicular stomatitis virus, Indiana serotype.
Reconstitution
Please refer to lot specific datasheet.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service