MABF2244
Anti-CD96 Antibody, clone 3.3

Synonym(s):
Cell surface antigen CD96, T cell-activated increased late expression protein, T-cell surface protein tactile
About This Item
biological source
rat
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
3.3, monoclonal
mol wt
calculated mol wt 67.13 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 100
technique(s)
flow cytometry: suitable
neutralization: suitable
isotype
IgG1
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
mouse ... Cd96(84544)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in mouse splenocytes.
Flow Cytometry Analysis: 1 µg from a representative lot detected CD96 in one million C57BL/6 mouse splenocytes.
Tested Applications
Flow Cytometry Analysis: A representative lot detected CD96 in Flow Cytometry application (Chan, C.J., et al. (2014). Nat Immunol. 15(5):431-8; Barrow, A.D., et al. (2018). Cell. 172(3):534-548.e19).
Neutralizing Assay: A representative lot detected CD96 in Neutralizing application (Chan, C.J., et al. (2014). Nat Immunol. 15(5):431-8; Barrow, A.D., et al. (2018). Cell. 172(3):534-548.e19).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service