Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2044

Sigma-Aldrich

Anti-MCPyV large T-antigen Antibody, clone CM2B4

clone CM2B4, from mouse

Synonym(s):

Large T antigen, MCV Large T antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CM2B4, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

shipped in

ambient

target post-translational modification

unmodified

General description

Merkel cells are neuroendocrine cells found in skin that have synaptic contacts with somatosensory afferents. These cells can turn malignant and form an aggressive form of skin cancer, which is known as Merkel cell carcinoma (MCC). A vast majority of MCC are caused by a polyomavirus known as Merkel cell polyomavirus (MCPyV or MCV). The MCPyV genome is reported to undergo clonal integration into the host cell chromosomes of MCC tumors and expresses small T antigen and truncated large T antigen. Full-length MCPyV large T-antigen is a 125 kDa nuclear protein, however, MCPyV T-antigens obtained from MCC have natural truncating mutations, which result in variably-sized, smaller proteins. Based on homology to other polyomaviruses, the MCPyV large and small T antigens are predicted to be oncogenic and contribute directly to the carcinogenesis of MCC. MCPyV large T antigen can serve as a specific marker for MCC.

Specificity

Clone CM2B4 is highly specific for MCPyV large T and 57kT isoforms, but does not detect MCPyV small T antigen.

Immunogen

KLH-conjugated linear peptide corresponding to RSRKPSSNASRGA sequence from the Merkel cell polyomavirus large T antigen exon 2 with a C-terminal cystiene.

Application

Anti-MCPyV large T-antigen, clone CM2B4, Cat. No. MABF2044, is a mouse monoclonal antibody that detects MCPyV large T antigen and has been tested for use in Immunohistochemistry, Immunoprecipitation, and Western Blotting for the detection of Merkel cell polyomavirus.
Western Blotting Analysis: A representative lot detected MCPyV large T-antigen in Western Blotting applications (Liu, X., et. al. (2011). J Biol Chem. 286(19):17079-90; Rodig, S.J., et. al. (2012) J Clin Invest. 122(12):4645-53).

Immunoprecipitation Analysis: A representative lot detected MCPyV large T-antigen in Immunoprecipitation applications (Liu, X., et. al. (2011). J Biol Chem. 286(19):17079-90).

Immunohistochemistry Analysis: A representative lot detected MCPyV large T-antigen in Immunohistochemistry applications (Rodig, S.J., et. al. (2012) J Clin Invest. 122(12):4645-53; Busam, K.J., et. al. (2009). Am J Surg Pathol. 33(9):1378-85).

Quality

Evaluated by Western Blotting in MKL-1 cell lysates.

Western Blotting Analysis: 1 µg/mL of this antibody detected MCPyV large T-antigen in 10 µg of MKL-1 cell lysates.

Target description

45 kDa observed for truncaed form. For intact form MW 125 kDaUncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service