Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1990

Sigma-Aldrich

Anti-Influenza A H5N1 Hemagglutinin Antibody, clone 9F4

clone 9F4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

9F4, monoclonal

Quality Level

isotype

IgG2bκ

Specificity

Clone 9F4 is a mouse monoclonal antibody that detects hemagglutinin from Influenza A virus. It targets an epitope within the vestigial esterase sub-domain.

Immunogen

Recombinant full-length hemagglutinin (HA) protein of Influenza A/chicken/Hatay/2004 H5N1 virus.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service