Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE2183

Sigma-Aldrich

Anti-MCM8 Antibody, clone 2D4

Synonym(s):

C20orf154, DNA helicase MCM8, Minichromosome maintenance 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D4, monoclonal

mol wt

calculated mol wt 93.69 kDa
observed mol wt ~95 kDa

purified by

using protein G

species reactivity

human, mouse

packaging

antibody small pack of 100

technique(s)

ChIP: suitable
electron microscopy: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... MCM8(84515)

Specificity

Clone 2D4 is a mouse monoclonal antibody that specifically detects MCM8 protein. It targets an epitope within 18 amino acids from the N-terminal half of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal half of human MCM8.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MCM8 in HeLa cell nuclear extract.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MCM8 in HEK293T and A-431 cell lysates.

Immunocytochemistry Analysis: A representative lot detected MCM8 in Immunocytochemistry applications (Kinoshita, Y., et al. (2008). Microsc Res Tech. 71(4):288-97).

Western Blotting Analysis: A representative lot detected MCM8 in Western Blotting applications (Johnson, E.M., et al. (2003). Nucleic Acids Res. 31(11):2915-25; Kinoshita, Y., et al. (2008). Microsc Res Tech. 71(4):288-97).

Electron Microscopy: A representative lot detected MCM8 in Electron Microscopy applications (Kinoshita, Y., et al. (2008). Microsc Res Tech. 71(4):288-97).

Immunoprecipitation Analysis: A representative lot immunoprecipitated MCM8 in Immunoprecipitation application (Kinoshita, Y., et al. (2008). Microsc Res Tech. 71(4):288-97).

Chromatin Immunoprecipitation Analysis (ChIP): A representative lot immunoprecipitated MCM8 in Chromatin Immunoprecipitation application (Kinoshita, Y., et al. (2008). Microsc Res Tech. 71(4):288-97).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

DNA helicase MCM8 (Uniprot; Q9UJA3; also known as Minichromosome maintenance 8) is encoded by MCM8 gene (Gene ID; 84515) in human. It is a nuclear protein, localizes to nuclear foci and to double-stranded DNA breaks. Its expression is highest in placenta, lung and pancreas and low in skeletal muscle and kidney. Also, expressed in various tumors with highest levels in colon and lung cancers. MCM8 is a member of MCM proteins, which are key components of the pre-replication complex, recruit replication-related proteins and form replication forks. It is a component of the MCM8-MCM9 complex which is involved in the repair of double-stranded DNA breaks (DBSs) and DNA interstrand cross-links (ICLs) by homologous recombination (HR). MCM8 is particularly important in the restart of stalled replication forks, contributing to the maintenance of genome stability. Additionally, it has been associated with meiotic recombination and DNA repair processes, highlighting its significance in preserving the integrity of the genetic material. The dysregulation of MCM8 has been linked to genomic instability and cancer development, emphasizing its role as a potential target for cancer therapy and as a biomarker for assessing genomic instability. (Ref.: Johnson, E.M., et al. (2003). Nucleic Acids Res. 31(11):2915-25. ; Kinoshita, Y., et al. (2008). Microsc Res Tech. 71(4):288-97).

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service