MAB5430-C
Anti-Tau Antibody, Cleaved (D421), clone Tau-C3, Ascites Free
clone Tau-C3, from mouse
Synonym(s):
Microtubule-associated protein tau, Tau, Cleaved (D421), Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau
Microtubule-associated protein tau, Asp421-cleaved N-terminal fragment, Neurofibrillary tangle protein, Asp421-cleaved
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
Tau-C3, monoclonal
species reactivity
rat, mouse, human
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)
General description
Specificity
Immunogen
Application
Immunofluorescence Analysis: A representative lots detected pathological tangles containing truncated Tau cleaved at D421 by fluorescent immunohistochemistry staining of Alzheimer′s diseased (AD) human brain sections (Guillozet-Bongaarts, A.L., et al. (2005). Neurobiol. Aging. 26(7):1015-1022; Horowitz, P.M., et al. (2004). J. Neurosci. 24(36):7895-7902).
Immunohistochemistry Analysis: A representative lot detected an increase of truncated Tau cleaved at D421 in the brain section from rTg4510 mice expressing human P301L mutant Tau transgene when compared with sections from non-transgenic mice (Ramalho, R.M., et al. (2008). Mol. Med. 14(5-6):309-317).
Immunohistochemistry Analysis: A representative lot immunostained neurofibrillary tangles (NFTs) and neuritic plaques (NPs) containing truncated Tau cleaved at D421 in frozen hippocampus sections from Alzheimer′s diseased (AD) human brains (Gamblin, T.C., et al. (2003). Proc. Natl. Acad. Sci. U. S. A. 100(17):10032-10037).
Western Blotting Analysis: A representative lot detected truncated Tau cleaved at D421 in frontal cortex and hippocampus tissue extracts from non-transgenic and rTg4510 transgenic mice with human P301L mutant Tau transgene (Ramalho, R.M., et al. (2008). Mol. Med. 14(5-6):309-317).
Western Blotting Analysis: A representative lots detected recombinantt Tau 1-421 fragment, but not 1-391 or 1-429 fragment or full-length Tau. Clone TauC3 detected tau fragment generated by caspase-mediated D421 cleavage of full-length recombinant Tau (Guillozet-Bongaarts, A.L., et al. (2005). Neurobiol. Aging. 26(7):1015-1022; Park, S.Y., and Ferreira, A. (2005). J. Neurosci. 25(22):5365-5375; Gamblin, T.C., et al. (2003). Proc. Natl. Acad. Sci. U. S. A. 100(17):10032-10037).
Immunocytochemistry Analysis: A representative lot detected a time-dependent increase of intracellular truncated Tau cleaved at D421 by fluorescent immunocytochemistry staining of paraformaldehyde-fixed primary embryonic rat cortical neurons subjected to fibrilar Aβ (1–42) treament (Gamblin, T.C., et al. (2003). Proc. Natl. Acad. Sci. U. S. A. 100(17):10032-10037).
Neuroscience
Neurodegenerative Diseases
Quality
Immunohistochemistry Analysis: An 1:250 dilution of this antibody detected Asp421-cleaved Tau in neurofibrillary tangles (NFTs) of Alzheimer′s diseased (AD) human brain tissue .
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service