MAB2638
Anti-Integrin beta7 Antibody, clone TN114
clone TN114, from hamster(Armenian)
Synonym(s):
integrin, beta 7, integrin beta 7 subunit, Gut homing receptor beta subunit
About This Item
Recommended Products
biological source
hamster (Armenian)
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
TN114, monoclonal
species reactivity
human
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ITGB7(3695)
General description
Specificity
Immunogen
Application
Cell Structure
Integrins
Quality
Flow Cytometry Analysis: 1 µg/mL of this antibody detected Integrin beta7 on human PBMC′s.
Target description
Physical form
Storage and Stability
Analysis Note
Human peripheral blood mononuclear cells (PBMC′s).
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service