Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GC0139

Sigma-Aldrich

Toluene & Ethanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352005

form

liquid

Quality Level

shelf life

12 mo. (from manufacture date)

color

APHA: ≤10

shipped in

ambient

storage temp.

no temp limit

Analysis Note

Expiration date12 months from mfg dateColor (APHA)10APHA maxDensity at 20°C0.8406-0.8606g/cm³FormClear liquid

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service