Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ECM505

Sigma-Aldrich

QCM Chemotaxis Cell Migration Assay, 24-well (3 µm), fluorimetric

Synonym(s):

Cell Migration Assay, Chemotaxis Assay Kit, cell migration assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.76

product line

Chemicon®

Quality Level

technique(s)

activity assay: suitable

Application

The QCM Chemotaxis 3 um 24-well Migration Assay is performed in a Migration Chamber, based on the Boyden chamber principle.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service