Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADCMXR0100LG2L

Mobius® ADC Reactor Single-Use Bag Assembly

new

Gold, 100 L, Ultimus® Film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

bag (Ultimus® bag)
polypropylene impeller(s)

Quality Level

Agency

meets EN 2.6.14
meets ISO 10993-4
meets JP 4.01
4.01

meets USP 661
meets USP 788
meets USP 85
meets USP 87
meets USP 88

sterility

sterile; irradiated (at 25-40 kGy)

product line

Mobius®

parameter

4-60 °C temp. range (39-140 °F)

capacity

100 L

application(s)

pharma/biopharma processes

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Mobius® ADC Reactor Single-Use Bag Assembly is made of Ultimus® Film technology, providing enhanced bag strength, improved durability, and leak resistance to ensure operator safety.
Incorporating woven nylon into the new film has increased its durability while maintaining the ease of handling, flexibility, and conformity that ADC processes demand.

Application

Specifically designed for the key conjugation step to manufacture Antibody Drug Conjugates (ADCs).

Packaging

Pack of 2 assemblies.

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTIMUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service