ABT70
Anti-WASH complex subunit 7 (SWIP) Antibody
from rabbit
Synonym(s):
WASH complex subunit 7
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunoprecipitation (IP): suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... WASHC4(23325)
General description
WASH complex subunit 7 (SWIP) is a component of the WASH complex belonging to the WASH S7 family. The WASH complex is localized at early endosomal subdomains involved in the activation and recruitment the ARP 2/3 complex in order to stimulate actin polymerization and tubulin interaction. Through its interaction with FAM21, WASH plays a regulatory role in the retromer-mediated trafficking of vesicles. It is also involved in the tubule fission, components required in endosomal sorting processes as transport intermediates.
Immunogen
GST-tagged recombinant protein corresponding to human WASH complex subunit 7.
Application
Detect WASH complex subunit 7 (SWIP) using this Anti-WASH complex subunit 7 (SWIP) Antibody validated for use in IP.
Research Category
Cell Structure
Cell Structure
Research Sub Category
Cytoskeletal Signaling
Cytoskeletal Signaling
Quality
Evaluated by Immunoprecipitation/Western Blot Analysis in HeLa immunoprecipitation with rabbit IgG and Anti-WASH complex subunit 7.
Immunoprecipitation/Western Blot Analysis: 5 µg/mL of this antibody detected WASH complex subunit 7 in 10 µg of HeLa immunoprecipitation with rabbit IgG and Anti-WASH complex subunit 7.
Immunoprecipitation/Western Blot Analysis: 5 µg/mL of this antibody detected WASH complex subunit 7 in 10 µg of HeLa immunoprecipitation with rabbit IgG and Anti-WASH complex subunit 7.
Target description
~135 kDa observed.
IgG heavy chain may be observed at approximately 50 kDa
IgG heavy chain may be observed at approximately 50 kDa
Physical form
Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
HeLa immunoprecipitation with rabbit IgG and Anti-WASH complex subunit 7
HeLa immunoprecipitation with rabbit IgG and Anti-WASH complex subunit 7
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service