Recommended Products
General description
GST-tagged recombinant protein corresponding to chicken MYPT1.
Quality
Evaluated by Western Blotting in HeLa cell lysate.
Western Blotting Analysis: A 1:200 dilution of this antibody detected MYPT1 in 10 µg of HeLa cell lysate.
Western Blotting Analysis: A 1:200 dilution of this antibody detected MYPT1 in 10 µg of HeLa cell lysate.
Physical properties
~130/140 kDa observed. Uncharacterized band(s) may appear in some lysates.
Physical form
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Other Notes
Human. Predicted to react with Chicken based on 100% sequence homology.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service