Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN895

Sigma-Aldrich

Anti-TDP-43 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

affinity purified immunoglobulin

Quality Level

clone

polyclonal

purified by

(Antigen Affinity Purified)

species reactivity

human, rabbit

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

General description

TDP43 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human TDP43.

Application

Epitope: Near C-terminus

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis (WB): 1 μg/mL of this antibody detected TDP-43 in HeLa cell lysate.

Physical properties

~30 and ~45 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service