Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN470

Sigma-Aldrich

Anti-EPAC1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Rap guanine nuclear exchange factor 3, RAPGEF3, cAMPGEF-I, bcm910, cAMP-GEFI, Exchange factor directly activated by cAMP, cAMP-regulated guanine nucleotide exchange factor I, EPAC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RAPGEF3(10411)

General description

Exchange protein directly activated by cAMP 1 (EPAC1) is also known as Rap guanine nuclear exchange factor 3 and cAMPGEF-I. EPAC1 is a guanine nucleotide exchange factor (GEF) for RAP1A and RAP2A small GTPases which are activated through cAMP binding. EPAC1 couples cAMP production to the activation of the Ras family GTPase, Rap. Activation of Rap drives several cellular processes such as integrin-mediated cell adhesion, vascular endothelial barrier formation, and cardiac myocyte gap junction formation. EPAC1 is part of a signaling complex in which it activates the PI3K gamma complex, which is involved in angiogenesis. EPAC1 may also regulate ion channel formation, intracellular Ca++ signaling, ion transporter activity, and exocytosis. EPAC1 is widely expressed, especially in brain, heart, kidney, pancreas, spleen, ovary, thyroid and spinal cord.

Immunogen

KLH-conjugated linear peptide corresponding to human EPAC1.

Application

Anti-EPAC1 Antibody is a highly specific rabbit polyclonal antibody, that targets Epac & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected EPAC1 in human kidney, thalamus, and cerebellum tissues.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in rat skeletal muscle tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected EPAC1 in 15 µg of rat skeletal muscle tissue lysate.

Target description

~103 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat skeletal muscle tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service