Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN347

Sigma-Aldrich

Anti-LGI4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich repeat LGI family member 4, LGI1-like protein 3, Leucine-rich glioma-inactivated protein 4, LGI4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LGI4(163175)

General description

The leucine-rich, glioma inactivated gene 4 (LGI4) is a member of the LGI family. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs). Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI4 have not been found to be associated with autodominant lateral temporal epilepsy (ADTLE). However, the LGI4 gene is located in a region linked to benign familial infantile convulsions and a GC-to-AT polymorphism in LGI4 was correlated with childhood absence epilepsy. Other studies showed that decreasing LGI4 expression in cultured cells inhibits myelination, indicating that LGI4 may play a role in neural development.

Specificity

Two isoforms of LGI4 are known to exist; this LGI4 antibody will recognize only the larger form. This LGI4 antibody is predicted to be specific to LGI4 and not recognize other LGI proteins.

Immunogen

KLH-conjugated linear peptide corresponding to human LGI4.

Application

Detect the LGI4 protein using this Anti-LGI4 validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in human brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected LGI4 in 15 µg of human brain tissue lysate.

Target description

~59 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS and 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service