Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE43

Sigma-Aldrich

Anti-Ebp1 NT Antibody

serum, from rabbit

Synonym(s):

Proliferation-associated protein 2G4, Cell cycle protein p38-2G4 homolog, hG4-1, ErbB3-binding protein 1, Cell cycle protein p38-2G4 homolog, ErbB-3 binding protein 1, ErbB3-binding protein Ebp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PA2G4(5036)

General description

Ebp1 is an RNA-binding protein involved in growth regulation. In cancer cells, this protein and has been implicated in growth inhibition and differentiation. Ebp1 is known to bind ribosomal and double-stranded RNA. It is a found in pre-ribosomal ribonucleoprotein complexes and is believed to play a role in ribosome assembly and rRNA processing. This protein is also a component of cytoplasmic bcl-2 messenger ribonucleoprotein. Through interactions with histion deacetylases, this protein can act transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes. Ebp1 also interacts with the cytoplasmic domain of the ErbB3 receptor, potentially transducing regulatory growth signals.

Specificity

Recognizes Ebp-1.

Immunogen

Epitope: N-terminus
Linear Peptide corresponding to the N-terminus of Ebp1.

Application

Anti-Ebp1 N-terminal Antibody is a Rabbit Polyclonal Antibody for detection of Ebp1 also known as Proliferation-associated protein 2G4 & has been validated in WB, ICC.
Immunocytochemistry:
A 1:500 dilution from a representative lot detected Ebp1 in NIH/3T3, A431, and HeLa cells.
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Chromatin Biology

Quality

Evaluated by Western Blotting in A431 cell lysate.

Target description

48 kDa Observed

Linkage

Replaces: 07-397

Physical form

Rabbit Polyclonal Serum with 0.05% sodium azide
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 Cell Lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew L Kraushar et al.
Molecular cell, 81(2), 304-322 (2020-12-29)
Protein synthesis must be finely tuned in the developing nervous system as the final essential step of gene expression. This study investigates the architecture of ribosomes from the neocortex during neurogenesis, revealing Ebp1 as a high-occupancy 60S peptide tunnel exit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service