Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE1439

Sigma-Aldrich

Anti-60S RPL5

from rabbit, purified by affinity chromatography

Synonym(s):

60S ribosomal protein L5, RPL5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

rabbit (based on 100% sequence homology), bovine (based on 100% sequence homology), horse (based on 100% sequence homology), porcine (based on 100% sequence homology), Xenopus (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RPL5(6125)

General description

60S ribosomal protein L5 (UniProt P46777; also known as RPL5) is encoded by the RPL5 (also known as DBA6, MSTP030) gene (Gene ID 6125) in human. Ribosomal proteins (RPs) are normally involved in the assembly of the translational machinery-ribosomes essential for protein production. RPL5, RPL11 and the 5S rRNA comprise the 5S ribonucleoprotein particle (RNP), an essential subcomplex of the large ribosomal subunit. RPL5 binds the 5S rRNA, the 5S rRNA/RPL5 complex then localizes to the nucleolus, where it binds RPL11 and is integrated into the ribosome. In response to ribosomal stress (RS) or nucleolar stress, the L5/L11/5S pre-ribosomal complex is redirected to inhibit MDM2 E3 ligase activity, thereby promoting p53 stabilization and p53-dependent cell cycle arrest. RPL5 and RPL11 can also bind TAp73 N-terminal TA domain in an RS-independent manner and thereby prevent MDM2 association with TAp73 target gene promoters, resulting in enhanced TAp73 transcriptional activity and TAp73-dependent apoptosis induction and cell cycle arrest.

Specificity

This rabbit polyclonal antibody targets a highly conserved sequence within the C-terminal half of 60S RPL5 from multiple species.

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence within the C-terminal half of human 60S RPL5.

Application

Anti-60S RPL5, Cat. No. ABE1439, is a highly specific rabbit polyclonal antibody that targets 60S ribosomal protein L5 and has been tested in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected 60S RPL5 in human (A431, HeLa, HepG2) and mouse (NIH/3T3) cells.

Western Blotting Analysis: 1 µg/mL from a representative lot detected 60S RPL5 in 10 µg of human (A431 epidermoid carcinoma & HUVEC) and rat (L6 myoblast) cell lysates.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected 60S RPL5 in 10 µg of HEK293 cell lysate.

Target description

~36 kDa observed. 34.23/34.27/34.33 kDa (human/mouse/rat) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service