AB9656
Anti-Tau phospho Serine 262 Antibody
Chemicon®, from rabbit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
phosphorylation (pSer262)
Gene Information
human ... MAPT(4137)
General description
Tau is a neuronal microtubule-associated protein found predominantly on axons and functions to promote tubulin polymerization and stabilize microtubules. Tau, in its hyperphosphorylated form, is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer′s disease (AD) brain. Hyperphosphorylated Tau is also found in neurofibrillary lesions in a range of other central nervous system disorders. Hyperphosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II (CK2), phosphorylate Tau. Serine 262 can be phosphorylated by GSK-3beta, PKA, CamKII, MARK and has been found to be a major site in AD brain.
Specificity
Tau phosphoSerine 262. The antibody recognizes Tau pSerine 262 in samples of recombinant human Tau treated with PKA for 45 minutes. The reactivity of the antibody is blocked with the pSerine 262 peptide but not the non-phosphopeptide or a generic phosphoSerine-containing peptide.
Immunogen
Synthetic peptide of amino acids surrounding the phosphoSerine 262 site of human Tau.
Application
Detect Tau phospho Serine 262 using this Anti-Tau phospho Serine 262 Antibody validated for use in WB.
Research Category
Neuroscience
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Neurodegenerative Diseases
Western blot: 1:1,000. Suggested blocking buffer is 5% BSA-TBST overnight at 2-8?C. Suggested antibody dilution buffer is 3% BSA-TBST. Suggested antibody incubation time is 2 hours at room temperature.
Optimal working dilutions must be determined by the end user.
Optimal working dilutions must be determined by the end user.
Storage and Stability
Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Fei Liu et al.
The Journal of biological chemistry, 280(3), 1790-1796 (2004-11-18)
Protein phosphatase (PP) 5 is highly expressed in the mammalian brain, but few physiological substrates have yet been identified. Here, we investigated the kinetics of dephosphoryation of phospho-tau by PP5 and found that PP5 had a K(m) of 8-13 microm
Fei Liu et al.
FEBS letters, 512(1-3), 101-106 (2002-02-20)
In Alzheimer's disease (AD) brain, microtubule-associated protein tau is abnormally modified by hyperphosphorylation and glycosylation, and is aggregated as neurofibrillary tangles of paired helical filaments. To investigate the role of tau glycosylation in neurofibrillary pathology, we isolated various pools of
Myriam Müller et al.
Journal of cell science, 123(Pt 2), 286-294 (2009-12-23)
Wee1 is well characterized as a cell-cycle checkpoint kinase that regulates the entry into mitosis in dividing cells. Here we identify a novel function of Wee1 in postmitotic neurons during the establishment of distinct axonal and dendritic compartments, which is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service