Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9358

Sigma-Aldrich

Anti-Neuropeptide Y2 Receptor Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NPY2R(4887)

Specificity

Recognizes Neuropeptide Y2 Receptor.

Immunogen

Synthetic peptide from Neuropeptide Y2 Receptor.

Application

Immunohistochemistry: 1:500-1:1,000 (see application notes that follow).

Optimal working dilutions must be determined by the end user.

APPLICATION NOTES FOR AB9358

IMMUNOHISTOCHEMISTRY

Suggested perfusion: 1) Ca2+-free Tyrode′s solution followed by 2) paraformaldehyde-picric acid fixative, and 3) 10% sucrose in PBS as a cryo-protectant. Tissues were rapidly dissected out and stored overnight in 0.1 M phosphate buffer (pH 7.4) containing 10% sucrose. Slide-mounted tissue sections were incubated with blocking buffer for 1 hour at room temperature. Primary antibody was diluted in blocking buffer to the appropriate working dilution. Blocking buffer was removed and the slides were then incubated at 2-8°C for 18-24 hours with AB9358. After rinsing in PBS 3 times sections were incubated for 60 minutes at room temperature with fluorescently labeled secondary antibody.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
This Anti-Neuropeptide Y2 Receptor Antibody is validated for use in IH for the detection of Neuropeptide Y2 Receptor.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service