Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5400

Sigma-Aldrich

Anti-GFR α3 Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GFRA3(2676)

Specificity

GFRa3. No reactivity with its homologues.

Immunogen

An eleven amino acid peptide corresponding to the N-terminus of the mouse GFR(3 protein.

Application

Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
This Anti-GFR α3 Antibody is validated for use in WB, IH for the detection of GFR α3.
Western blot: 1:500

Immunohistochemistry: 1:500-1:1,000

Optimal working dilutions must be determined by end user.

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50 μL of sterile distilled water. Centrifuge if necessary to remove any residue.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C for up to 6 months after date of receipt. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service