Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

9030

Sigma-Aldrich

Glucagon

Iodinated Glucagon -HPLC purified & prepared fresh monthly.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
form:
solid

label

125Iodine labeled

Quality Level

form

solid

NCBI accession no.

UniProt accession no.

shipped in

ambient

Gene Information

human ... GCG(2641)

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service