Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902266

Supelco

Rapid-action connecting nipple

3/8 in. (gas side)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

fitting

3/8 in. (gas side)

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

(gas side) with 3/8" thread
Our withdrawal and safety components are easily interconnectable thanks to a wide selection of reducers, adapters and couplings. Due to the wide variety, many different customized installations are possible – everything for your individual requirements.
This rapid-action connector is made of stainless steel with a special high quality sealing for maximum protection of our solvents quality. It is equipped with nipple (male) Type 21 and G3/8 outer thread.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service