8.52405
Fmoc-Glu(OtBu)-(Dmb)Gly-OH
Novabiochem®
Synonym(s):
Fmoc-Glu(OtBu)-(Dmb)Gly-OH
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C35H40N2O9
Molecular Weight:
632.70
UNSPSC Code:
12352209
NACRES:
NA.22
Recommended Products
Quality Level
product line
Novabiochem®
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
carboxylic acid
storage temp.
2-8°C
Related Categories
General description
Fmoc-Aaa-(Dmb)Gly-OH dipeptides offer the same benefits as pseudoproline dipeptides such as prevention of aggregation and improvements in product quality but for peptide sequences containing Gly. They are extremely easy to use. Standard coupling methods like PyBOP®/DIPEA or DIPCDI/HOBt can be used for their introduction. Removal of the Dmb group and regeneration of the glycine residue occurs during the course of standard TFA-mediated cleavage reaction.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aspartimide Formation in Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aspartimide Formation in Fmoc SPPS
Analysis Note
Color (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -31.0 - -27.0 °
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -31.0 - -27.0 °
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service