Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.19145

Sigma-Aldrich

Chlorobenzene / Acetic acid (mixture 2:1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601
NACRES:
NA.22

Quality Level

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F - (calculated)

Flash Point(C)

26 °C - (calculated)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service