565763
γ-Secretase Inhibitor VI
The γ-Secretase Inhibitor VI, also referenced under CAS 287196-57-0, controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.
Synonym(s):
γ-Secretase Inhibitor VI, 1-(S)- endo-N-(1,3,3)-Trimethylbicyclo[2.2.1]hept-2-yl)-4-fluorophenyl Sulfonamide, 1-(S)-endo-N-(1,3,3)-Trimethylbicyclo[2.2.1]hept-2-yl)-4-fluorophenyl Sulfonamide
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
DMSO: 200 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
Biochem/physiol Actions
Aβ₄₂ production
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service