Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5501

SAFC

CellPrime® rLysozyme recombinant human lysozyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32011401

material

off-white

Quality Level

type

for cell culture

Assay

≥75% (Protein Content (A280 0.1%=2.46))
≥85% (SDS-PAGE)

form

powder (

)

specific activity

>100,000 units/mg protein

shelf life

24 mo. (at 2–8 °C. More studies are underway.)

purified by

using Protein A

loss

<10% loss on drying (w/w)

solubility

soluble (Clear solution at 10 mg/ml in phosphate buffered saline, pH 7.4 or H<sub>2</sub>O)

General description

CellPrime® rLysozyme is a non-animal origin recombinant human lysozyme specifically manufactured under cGMP standards for use in biopharmaceutical production processes.

Physical form

A lyophilized, off-white powder

Storage and Stability

Shipping and storage at 2–8 °C

Legal Information

CellPrime is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For research and further manufacturing use only. Not for use in humans or for clinical diagnostic applications.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service