5.34352
MNK Inhibitor IV, Mnk-I1
Synonym(s):
MNK Inhibitor IV, Mnk-I1, MNK1 Inhibitor, Mnk-I1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H27F2N5O2S
Molecular Weight:
463.54
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
light beige
solubility
DMSO: 25 mg/mL
storage temp.
2-8°C
General description
A cell-permeable thieno[2,3-d]pyrimidine compound that acts as a selective inhibitor of MAP kinase-interacting kinases 1 and 2 (Mnk1 and 2: IC50 = 23 and 16 nM for MNK1 and MNK2, respectively). Displays much reduced activity against several other protein kinases and does not affect the phosphorylation of ERK, Akt, mTORC1, and ribosomal S6 kinase. Shown to strongly inhibit eIF4E phosphorylation (~1 µM) and abolished the activity completely at higher concentration in MDA-MB-231 and SSC25 cancer cell (~ 1 µM). Shown to be much more effective Mnk inhibitor than CGP57380 (Cat. No. 454861). Reported to inhibit the migration of mouse embryonic fibroblasts and MDA-MB-231 and SCC25 cells, but does not affect their viability and proliferation.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
MNK
MNK
Reversible: yes
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Beggs, J.E., et al. 2015. Biochem. J.467, 63.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service