Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33514

Sigma-Aldrich

GADD45b/MKK7 Inhibitor, DTP3

Synonym(s):

GADD45b/MKK7 Inhibitor, DTP3, (R)-2-((R)-2-acetamido-3-(4-hydroxyphenyl)propanamido)-N-((R)-1-amino-1-oxo-3-phenylpropan-2-yl)-5-guanidinopentanamide trifluoroacetate, GADD45b-MKK blocker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35N7O5 · xC2HF3O2
Molecular Weight:
525.60 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

65 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 100 mg/mL
water: 100 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)NC(CCCN=C(N)N)C(=O)NC(CC2=CC=CC=C2)C(=O)N

InChI key

AOUZPXZGMZUQQS-UHFFFAOYSA-N

General description

A cell permeable, bioavailable D-tripeptide that specifically binds to MKK7 in 1:1 stoichiometry and disrupts the GADD45b/MKK7 complex in an allosteric mechanism (Kd = 65 nM, IC50 = 160 pM). Does not affect the activity of 142 other protein kinases. Exhibits potent and selective activity in both multiple myeloma (MM) and non-MM cell lines expressing high levels of GADD45b, however, it is completely inactive in tumor cells featuring low GADDb expression. Displays potency similar to bortezomib, but has about 100-fold greater selectivity for cancer cells, and unlike bortezomib, it does not display any toxic effects in normal cell lines. Shown to activate JNK, but does not affect p38, ERK, or IKK/NF-κB signaling in sensitive GADD45b dependent MM cell lines. Abolishes the growth of myeloma xenografts in mice (14.5 mg/kg/day) and retains anticancer activity in an orthotopic xenograft model of MM. Displays desirable pharmacokinetic profile (t1/2 = 1.26 h and AUC = 6.43 mg/h/ml; at ~10 mg/kg, i.v).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GADD45b/MKK7
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-YRF-NH2

Physical form

Supplied as a HCl salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tomatore, L., et al. 2014. Cancer Cell.26, 495.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service