Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

513040-M

Millipore

PD 174265

A potent, cell-permeable, reversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC₅₀ = 450 pM).

Synonym(s):

PD 174265, 4-[(3-Bromophenyl)amino]-6-propionylamidoquinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15BrN4O
CAS Number:
Molecular Weight:
371.23
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 200 mg/mL

storage temp.

−20°C

SMILES string

Brc1cc(ccc1)Nc2ncnc3c2cc(cc3)NC(=O)CC

InChI

1S/C17H15BrN4O/c1-2-16(23)21-13-6-7-15-14(9-13)17(20-10-19-15)22-12-5-3-4-11(18)8-12/h3-10H,2H2,1H3,(H,21,23)(H,19,20,22)

InChI key

WUPUZEMRHDROEO-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50 = 450 pM). The reversible nature of this compound makes it a less effective antitumor agent than PD 168393 (Cat. No. 513033).

Biochem/physiol Actions

Primary Target
EGFR tyrosine kinase activity
Target IC50: 450 pM against EGF receptor (EGFR) tyrosine kinase activity

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fry, D.W., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 12022.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

495.3 °F

Flash Point(C)

257.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service