Recommended Products
Assay
≥96% (for each compound, HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
solubility
DMSO: 50 mg/mL
storage temp.
2-8°C
General description
This set contains two distinct calcium channel blockers acting through two different mechanisms.
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Physical form
A set consists of 10 mg each of CRAC Channel Inhibitor III, PYR6 (Cat. No. 5.09710.0001) and TPRC3 Channel Inhibitor III, PYR10 (Cat. No. 5.09711.0001).
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Schleifer, H., et al. 2012. Br. J. Pharmacol. 167, 1712.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service