Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09584

Sigma-Aldrich

ISR Inhibitor, ISRIB

Synonym(s):

ISR Inhibitor, ISRIB, trans-2-(4-Chlorophenoxy)-N-(4-(2-(4-chlorophenoxy)acetylamino)cyclohexyl)acetamide, Integrated Stress Response Inhibitor, UPR Inhibitor, eIF2α (pS51) Signaling Inhibitor, eIF2S1 (pS51) Signaling Inhibitor, eIF2α (pS51) Signaling Inhibitor, eIF2S1 (pS51) Signaling Inhibitor, trans-2-(4-Chlorophenoxy)-N-(4-(2-(4-chlorophenoxy)acetylamino)cyclohexyl)acetamide, Integrated Stress Response Inhibitor, UPR Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24Cl2N2O4
CAS Number:
Molecular Weight:
451.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥93% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

C1CC(CCC1NC(=O)COC2=CC=C(C=C2)Cl)NC(=O)COC3=CC=C(C=C3)Cl

InChI

1S/C22H24Cl2N2O4/c23-15-1-9-19(10-2-15)29-13-21(27)25-17-5-7-18(8-6-17)26-22(28)14-30-20-11-3-16(24)4-12-20/h1-4,9-12,17-18H,5-8,13-14H2,(H,25,27)(H,26,28)

InChI key

HJGMCDHQPXTGAV-UHFFFAOYSA-N

General description

The cell-permeable trans-diastereomer of a bis-glycolamide compound that acts as a potent and selective blocker ofeIF2α/eIF2S1-Ser51 phosphorylation-initiated unfold protein response/UPR (Effec. conc. 25 to 200 nM in HEK293T, HeLa, MEF, U2OS cultures) upon ER stress induction by Cys/Met starvation, HRI activation (6 µM KM09748), Thapsigargin/Tg (100 to 500 nM; 586006 ) or Tunicamycin/Tm (2 to 5 µg/mL; Cat. No. 654380) treatment without affecting UPR responses independent of eIF2α (pS51), including ATF6 cleavage and IRE1-initiated XBP1s production. Although noncytotoxic in the absence of ER stress, ISRIB does prevent URP from properly restoring ER homeostasis following ER stress induction, causing enhanced apoptotic cell death in Tg- or Tm-treated HEK293T (2 µg/mL Tm & 200 nM ISRIB) and HeLa (25 nM ISRIB and 5 µg/mL Tm or 500 nM Tg) cultures. Reported to exhibit good pharmacokinetics with blood-brain permeability (plasma/brain Tmax ≈ 6/6 h; Cmax ≈ 180/100 ng/mL; C24 h ≈ 55/33 ng/mL; 2.5 mg/kg i.p.) in mice and significantly enhance hippocampus-dependent learning of mice subjected to Morris water maze (0.25 mg/kg/d i.p.) in vivo. Likewise, single amygdala administration via cannulation (25 ng/0.5 µL/275-350 g rat) immediately following auditory fear conditioning significantly enhances long-term, but not short-term, post-conditioning auditory response (% response above basal at 4 h/24 post conditioning = 27/33 vs. 27/22 among vehicle-treated control rats) in rats in vivo. Although the exact molecular target is not yet established, ISRIB most likely exerts its effect by modulating the interaction between the guanine nucleotide exchange factor eIF2B and the phosphorylated eIF2α (pS51).

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Sidrauski, C., et al. 2013 Elife2, e00498.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service