Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496915

Sigma-Aldrich

Oridonin, R. rubescens

A cell-permeable diterpenoid compound that possesses anti NF-κB activity and displays antiproliferative and antiangiogenic properties.

Synonym(s):

Oridonin, R. rubescens

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O6
CAS Number:
Molecular Weight:
364.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

RTECS - NZ8177000

Assay

≥93% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O1[C@]2([C@]43[C@H]([C@]5([C@H]([C@@H]2O)C(CC[C@@H]5O)(C)C)C1)CC[C@H]([C@H]4O)C(=C)C3=O)O

InChI

1S/C20H28O6/c1-9-10-4-5-11-18-8-26-20(25,19(11,14(9)22)15(10)23)16(24)13(18)17(2,3)7-6-12(18)21/h10-13,15-16,21,23-25H,1,4-8H2,2-3H3/t10-,11-,12-,13+,15+,16-,18+,19-,20+/m0/s1

InChI key

SDHTXBWLVGWJFT-XKCURVIJSA-N

General description

A cell-permeable diterpenoid compound that possesses anti NF-κB activity and displays antiproliferative (ED50 = ~ 2.7 µg/ml in lymphoid malignant cells) and antiangiogenic properties (significantly inhibits network formation of HMEC-1 cells at 2.5 µg/ml). Reported to affect DNA synthesis, induce apoptosis and initiate cell cycle arrest. Shown to efficiently block both TNF-α and LPS-induced NF-κB activity in Jurkat and in RAW264.7 murine macrophages, and inhibit p65 NF-κB transcriptional activity (IC50 = ~ 5 µg/ml in MT-1 cells) by disrupting NF-κB DNA-binding activity without interfering with its nuclear translocation.
A cell-permeable inhibitor of NF-κB activity that displays antiproliferative (ED50 ~ 2.7 µg/ml in lymphoid malignant cells) and antiangiogenic properties (significantly inhibits network formation of HMEC-1 cells at 2.5 µg/ml). Reported to affect DNA synthesis, induce apoptosis, and initiate cell cycle arrest. Shown to efficiently block both TNF-α and LPS-induced NF-κB activity in Jurkat and in RAW264.7 murine macrophages. Also inhibits p65 NF-κB transcriptional activity (IC50 ~5 µg/ml in MT-1 cells) by disrupting NF-κB DNA-binding activity without interfering with its nuclear translocation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NF-κB transcriptional activity
Product does not compete with ATP.
Reversible: no
Target IC50: 5 µg/ml inhibiting p65 NF-κB transcriptional activity in MT-1 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Ikezoe, T., et al. 2005. Mol. Cancer Ther.4, 578.
Meade-Tollin, L.C., et al. 2004. J. Nat. Prod.67, 2.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service