Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475958

Sigma-Aldrich

c-Raf Inhibitor

The c-Raf Inhibitor controls the biological activity of c-Raf. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

c-Raf Inhibitor, 1-(3-(1,4-dihydroimidazo[4,5-c]pyrazol-5-yl)-4-methylphenyl)-3-(3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl)urea, c-Raf Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19F3N8O
Molecular Weight:
480.45
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable pyrazoloimidazole phenyl urea derivative that acts as a selective type II inhibitor of c-Raf. It shows an IC50 value of 50 nM in an enzymatic assay, and comparable anti-proliferative activities to sorafenib against A375P (GI 50 = 2.68 µM) and WM3629 (GI50 = 0.86 µM) human melanoma cell lines. When screening against a panel of 30 different kinases at a concentration of 10 µM, it exerts an inhibitory activity of 96% on c-Raf, but below 50% for most of the other kinases.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yu, H., et al. 2010. Bioorg. Med. Chem. Lett. 20, 3805.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service