Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

412511

Sigma-Aldrich

IRE1 Inhibitor II

The IRE1 Inhibitor II controls the biological activity of IRE1.

Synonym(s):

IRE1 Inhibitor II, 3ʹ-Formyl-4ʹ-hydroxy-5ʹ-methoxybiphenyl-3-carboxamide, ER-to-Nucleus Signaling 1 Inhibitor II, Inositol-Requiring Protein 1 Inhibitor II, ER-to-Nucleus Signaling 1 Inhibitor II, Inositol-Requiring Protein 1 Inhibitor II, 3ʹ-Formyl-4ʹ-hydroxy-5ʹ-methoxybiphenyl-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO4
CAS Number:
Molecular Weight:
271.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

NC(C1=CC=CC(C2=CC(C([H])=O)=C(O)C(OC)=C2)=C1)=O

InChI key

CVOMLTZPVVYGAP-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service