411483
Mouse Anti-Human IgG₃, Hinge Specific (HP6047) Biotin Conjugate
liquid, clone HP6047, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.77
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
secondary antibodies
clone
HP6047, monoclonal
form
liquid
does not contain
preservative
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG3
shipped in
ambient
storage temp.
−20°C
target post-translational modification
unmodified
Related Categories
General description
Purified mouse monoclonal antibody conjugated to biotin. Recognizes human IgG3 hinge region.
This Mouse Anti-Human IgG₃, Hinge Specific (HP6047) Biotin Conjugate is validated for use in ELISA for the detection of Human IgG₃, Hinge Specific.
Immunogen
Human
Application
ELISA (1:500-1:2000, see comments)
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
In PBS, 1% BSA, pH 7.4.
Reconstitution
Following initial thaw, aliquot and freeze (-20°C).
Other Notes
Biotinylated with hydroxysuccinimide ester biotin. This biotin-labeled antibody is an extremely versatile reagent because of the high binding affinity for avidin and avidin-labeled compounds. Variables associated with assay conditions will dictate the proper working dilution.
Cross-reactivity by ELISA against human myeloma proteins:
Human IgG1: ≤0.1%
Human IgG2: ≤1%
Human IgG3: 100%
Human IgG4: ≤0.1%
Human IgA: ≤0.1%
Human IgE: ≤0.1%
Human IgM: ≤0.1%
Cross-reactivity by ELISA against chimeric antibodies:
Human IgG1: <0.001%
Human IgG2: <0.001%
Human IgG3: 100%
Human IgG4: <0.001%
Human IgA: <0.001%
Human IgE: <0.001%
Human IgM: <0.001%
Cross-reactivity by ELISA against human myeloma proteins:
Human IgG1: ≤0.1%
Human IgG2: ≤1%
Human IgG3: 100%
Human IgG4: ≤0.1%
Human IgA: ≤0.1%
Human IgE: ≤0.1%
Human IgM: ≤0.1%
Cross-reactivity by ELISA against chimeric antibodies:
Human IgG1: <0.001%
Human IgG2: <0.001%
Human IgG3: 100%
Human IgG4: <0.001%
Human IgA: <0.001%
Human IgE: <0.001%
Human IgM: <0.001%
Hamilton, R.G. 1990. Ann. Biol. Clin.48, 473.
Fasullo, F.J., et al. 1989. Clin. Chem.35, 364.
Hamilton, R.G. 1987. Clin. Chem.33, 1707.
Jefferis, R., et al. 1986. Immunol. Letters10, 223.
Fasullo, F.J., et al. 1989. Clin. Chem.35, 364.
Hamilton, R.G. 1987. Clin. Chem.33, 1707.
Jefferis, R., et al. 1986. Immunol. Letters10, 223.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service